Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPP4B Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341167 Supplier Novus Biologicals Supplier No. NBP282096MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INPP4B Polyclonal antibody specifically detects INPP4B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry

Specifications

Antigen INPP4B
Applications Western Blot, ELISA, Immunocytochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias inositol polyphosphate-4-phosphatase, type II, 105kDa, type II inositol-3,4-bisphosphate 4-phosphatase
Host Species Rabbit
Immunogen INPP4B antibody was raised against a 17 amino acid peptide near the amino terminus of human INPP4B. The immunogen is located within the last 50 amino acids of INPP4B. Amino Acid Squence: QFTSIQKTPNEPQLEF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 8821
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.