Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP5J Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109861 Supplier Invitrogen™ Supplier No. PA5109861
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTWNVGTAMP PDDVTSLLHL GGGDDSDGAD MIAIGLQEVN SMLNKRLKDA LFTDQWSELF MDALGPFNFV LVSSVRMQGV ILLLFAKYYH LPFLRDVQTD CTRTGLGGYW GNKGGVSVRL AAFGHMLCFL NCHLPAHMDK AEQRKDNFQT ILSLQQFQGP GAQGILDHDL VFWFGDLNFR IESYDLHFVK FAIDSDQLHQ LWEKDQLNMA KNTWPILKGF QEGPLNFAPT FKFDVGTNKY DTSAKKRKPA WTDRILWKVK APGGGPSPSG RKSHRLQVTQ HSYRSHMEYT VSDHKPVAAQ F.

Inositol and phosphatidylinositol phosphates are important for numerous cellular processes, including neuronal survival and signal transductions from growth factors, neurotransmitters and G protein coupled receptors. INPP5J (inositol polyphosphate-5-phosphatase J), also known as phosphatidylinositol 4,5-bisphosphate 5-phosphatase A, PIPP (proline-rich inositol polyphosphate 5-phosphatase), PIB5PA or INPP5, is a 1,006 amino acid cytoplasmic protein that assists in the conversion of inositol 1,4-bisphosphate from inositol 1,4,5-trisphosphate. Expressed in lung, stomach, kidney, brain, small intestine and heart, INPP5J localizes to membrane ruffles, where it may also participate in modulating inositol and phosphatidylinositol polyphosphate-binding proteins. Encoded by a gene located on human chromosome 22, INPP5J undergoes alternative splicing to produce three isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5J
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INPP5J
Gene Accession No. P59644, Q15735, Q9JMC1
Gene Alias Inositol polyphosphate 5-phosphatase J; inositol polyphosphate-5-phosphatase J; Inpp; INPP5; Inpp5j; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; phosphatidylinositol 4,5-bisphosphate 5-phosphatase A; Pib5pa; Pipp; Proline-rich inositol polyphosphate 5-phosphatase
Gene Symbols INPP5J
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-360 of human INPP5J (NP_001002837.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170835, 171088, 27124
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.