Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSIG-1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. N11055244UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11055244UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N11055244UV Supplier Novus Biologicals Supplier No. NB11055244UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen INSIG-1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405
Gene Symbols INSIG1
Host Species Rabbit
Immunogen A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-ProtNo. O15503.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3638
Test Specificity This is specific for Insig1 but not Insig2.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.