Learn More
Description
Specifications
Specifications
| Antigen | INSIG-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human INSIG-1 (NP_005533.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
