Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ InSight Accessories

Maximize your Thermo Scientific™ InSight with essential accessories.

Supplier:  Thermo Scientific™ ACCCON1016

 View more versions of this product

Catalog No. 11-676-506


Only null left
Add to Cart

Description

Description

Thermo Scientific™ InSight accessories include replacement batteries, mote cable assembly, terminal block kits, power cables and more.

  • Replacement C battery Li ion
  • Mote commissioning cable assembly
  • External PSU 110V for T3C1D2 mote
  • Terminal block kit
  • InSightLink gateway power cable
  • InSightLink gateway
  • Internal 12V power cable for T3C1D2 mote
TRUSTED_SUSTAINABILITY
Specifications

Specifications

InSightLink Gateway Power Cable
InSightLink gateway power cable
Cold Storage
Mote Commissioning Cable Assembly
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

Not available outside North America