Learn More
Thermo Scientific™ InSight Sensors
Thermo Scientific™ InSight sensors work with the mote transmitters, relaying data to the InSight Cloud service.
Supplier:  Thermo Scientific™ SENSRTD1002
 View more versions of this product
Catalog No. 11-676-533
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Thermo Scientific™ InSight sensors relay data from the motes on your wireless account to the InSight Cloud service. In the event of internet failure, Insight Cloud will notify IT and the InSight link will log sensor data for data backfill on recovery.
  • Door access sensor
  • RTD sensor
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RTD Assembly
RTD: RE style RTD assembly; single wire-wound 100 ohm platinum element, Class B tolerance; 14 foot PFA
Remote Monitoring, Asset Management Solution
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Not available outside North America