Learn More
Thermo Scientific™ InSight Sensors
Thermo Scientific™ InSight sensors work with the mote transmitters, relaying data to the InSight Cloud service.
$81.20 - $4330.00
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
11-676-405
|
Thermo Scientific™
INSPTCO2A001 |
CO2 sensor USB (spare part) |
Each for $4,330.00
|
|
|||||
11-676-400
|
Thermo Scientific™
INSPTCURT001 |
Each for $132.40
|
|
||||||
11-676-401
|
Thermo Scientific™
INSPTCURT002 |
CT 0-80amp (spare part) |
Each for $136.00
|
|
|||||
11-676-403
|
Thermo Scientific™
INSPTDOOR001 |
Each for $81.20
|
|
||||||
11-676-406
|
Thermo Scientific™
INSPTMRHI002 |
Temp/RH sensor (spare part) |
Each for $2,710.00
|
|
|||||
11-676-399
|
Thermo Scientific™
INSPTSMRT001 |
RTD Smart Cable (spare part) |
Each for $126.00
|
|
|||||
11-676-402
|
Thermo Scientific™
INSPTSMRT002 |
CT Smart Cable (spare part) |
Each for $146.70
|
|
|||||
11-676-398
|
Thermo Scientific™
INSPTTEMP001 |
Each for $440.50
|
|
||||||
11-676-533
|
Thermo Scientific™
SENSRTD1002 |
RTD: RE style RTD assembly; single wire-wound 100 ohm platinum element, Class B tolerance; 14 foot PFA | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
11-676-404
|
Thermo Scientific™
INSPTHUMI001 |
Temp/RH (<90% humidity) sensor (spare part) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Thermo Scientific™ InSight sensors relay data from the motes on your wireless account to the InSight Cloud service. In the event of internet failure, Insight Cloud will notify IT and the InSight link will log sensor data for data backfill on recovery.
- Door access sensor
- RTD sensor
- Warranty varies by country, please contact us for details
Not available outside North America
Your input is important to us. Please complete this form to provide feedback related to the content on this product.