Learn More
Description
Specifications
Specifications
| Antigen | INSL5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | insulin-like 5, Insulin-like peptide 5, insulin-like peptide INSL5, MGC126695, MGC126697, PRO182, UNQ156 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human INSL5 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
