Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSM1 Antibody (INSM1/6286R), HRP, Novus Biologicals™
SDP

Catalog No. NB179795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179795 Supplier Novus Biologicals Supplier No. NBP314105H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen INSM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone INSM1/6286R
Conjugate HRP
Formulation PBS
Gene Alias IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1
Host Species Rabbit
Immunogen Recombinant fragment (around aa81-125) corresponding to the N-terminus of human INSM1 (Uniprot: Q01101 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3642
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.