Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSM1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358480 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358480 Supplier Novus Biologicals Supplier No. NBP176307MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INSM1 Polyclonal antibody specifically detects INSM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen INSM1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human IA-1. The immunogen is located within amino acids 90 - 140 of IA-1. Amino Acid Squence: YSPTRPVSREHEKHKYFER
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3642
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.