Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSM2 Antibody (INSM2/4291), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB386231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386231 Supplier Novus Biologicals Supplier No. NBP308530UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

INSM2 Monoclonal antibody specifically detects INSM2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen INSM2
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone INSM2/4291
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias insulinoma-associated 2, Nbla106, zinc finger protein IA-6
Host Species Mouse
Immunogen Recombinant fragment of human INSM2 protein (around aa 432-540) (exact sequence is proprietary) (Uniprot: Q96T92 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84684
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.