Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSRR/IR-related receptor Antibody (326929), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348557 Supplier Novus Biologicals Supplier No. FAB3055MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

INSRR/IR-related receptor Monoclonal specifically detects INSRR/IR-related receptor in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen INSRR/IR-related receptor
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 326929
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.1, insulin receptor-related protein, insulin receptor-related receptor, IRRIR-related receptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human INSRR, Leu24-Leu923, Accession # P14616
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3645
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.