Learn More
Novus Biologicals™ INSTA-Blot Breast Tissue OncoPair
Click to view available options
Quantity:
1 Membrane
Description
OncoPair INSTA-Blots™ are ready-to-use PVDF western blot membranes containing denatured protein lysates. The NBP2-30333 OncoPair Breast Set contains five ready-to-use Breast OncoPair INSTA-Blots™. The donor/patient tissues in this set collectively represent a range of breast tumor types, stages and patient ages. The blots are themed by breast cancer type or stage, each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors. There are 34 different breast cancer patient donors represented in the five Blot Set, with one patient donor repeated. Store at room temperature.
Specifications
Specifications
Description | INSTA-Blot Breast Tissue OncoPair |
For Use With (Application) | Western Blot |
Quantity | 1 Membrane |