Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ INSTA-Blot Colon Tissue OncoPair
SDP

Catalog No. NBP230129 Shop All Novus Biologicals Products
Click to view available options
Quantity:
1 Membrane

Applications: Western Blot

  • Proteomic discovery tool
  • Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
  • Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
  • Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
  • Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
  • Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
  • Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines

Recommended Storage

Store at room temperature

Specifications

For Use With (Application) Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors
Quantity 1 Membrane
Material PVDF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.