Learn More
Description
- Proteomic discovery tool
- Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
- Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
- Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
- Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
- Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
- Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Recommended Storage
Store at room temperature
Specifications
Specifications
| For Use With (Application) | Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors |
| Quantity | 1 Membrane |
| Material | PVDF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
