Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ INSTA-Blot Mouse Brain
SDP

Catalog No. NBP230120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Membrane
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230120 1 Membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230120 Supplier Novus Biologicals™ Supplier No. NBP230120
Add to Cart
Add to Cart

PVDF membrane containing denatured protein from cells or tissue lysates

  • Screening proteins from various human, mouse and rat tissues
  • Contains mouse brain lysate (30μg total protein per lane) on 4-20% Tris-Glycine mini gel
  • Prepared from resolution by SDS-PAGE at 125V for 1 hour
  • Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
  • Stained with amido black for visualization of proteins

Recommended Storage

Store at room temperature

Specifications

For Use With (Application) Screening proteins from various human, mouse and rat tissues
Quantity 1 Membrane
Material PVDF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.