Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ INSTA-Blot Ovary Tissue OncoPair

Click to view available options
Quantity:
1 Membrane
Description
- Proteomic discovery tool
- Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
- Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
- Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
- Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
- Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
- Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Recommended Storage
Store at room temperature
Specifications
Specifications
For Use With (Application) | Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors |
Quantity | 1 Membrane |
Material | PVDF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction