Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Antibody (182410) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB400067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400067 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400067 Supplier Novus Biologicals Supplier No. FAB1417Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Bovine samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen Insulin
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 182410
Conjugate DyLight 755
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Rat
Immunogen E. coli-derived recombinant human Insulin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Mouse, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.