Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Antibody (E2-E3 + 2D11-H5 (same as INS04 + INS05)), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP234612X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234612X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234612X Supplier Novus Biologicals Supplier No. NBP234612AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin Monoclonal specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Insulin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone E2-E3 + 2D11-H5 (same as INS04 + INS05)
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Purified pig insulin, conjugated to KLH (E2-E3 & 2D11-H5) (Uniprot: P01308)
Molecular Weight of Antigen 6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.
Target Species Human, Mouse, Pig, Bovine, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.