Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Antibody (IN-05) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119712 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119712 Supplier Novus Biologicals Supplier No. NBP309089
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin Monoclonal specifically detects Insulin in Human, Porcine, Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Radioimmunoassay.

Specifications

Antigen Insulin
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Functional Assay, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IN-05
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Radioimmunoassay
Formulation PBS, 0.2 um filter sterilized
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Mouse
Immunogen Porcine insulin.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Pig, Bovine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.