Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Antibody (IRDN/1980R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB390209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390209 Supplier Novus Biologicals Supplier No. NBP308836AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Insulin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IRDN/1980R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Rabbit
Immunogen Recombinant human full-length Insulin protein (IRDN) (Uniprot: P01308)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Mouse, Pig, Bovine, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.