Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Antibody (rIRDN/4782), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32052120UG
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Insulin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Recombinant full-length human Insulin protein | |
20 μg | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
3630 | |
Store at 4&drg;C. Do not freeze. | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
rIRDN/4782 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction