Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Antibody (rIRDN/4782), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $454.00
Specifications
Antigen | Insulin |
---|---|
Clone | rIRDN/4782 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.1-0.2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Insulin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
10mM PBS with 0.05% BSA | |
3630 | |
IgG2a κ | |
Protein A or G purified |
rIRDN/4782 | |
Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Recombinant full-length human Insulin protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title