Learn More
Description
Specifications
Specifications
| Antigen | Insulin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM531 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin |
| Gene Symbols | INS |
| Host Species | Mouse |
| Immunogen | Full length (1-84 amino acid) Purified pig insulin, conjugated to KLH (Uniprot: P01308) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
