Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Mouse anti-Human, Mouse, Clone: 4B6A7, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 661981IG150UL
Description
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.Specifications
| Insulin | |
| Monoclonal | |
| 1.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01308 | |
| INS | |
| Insulin Fusion Protein Ag8630 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 4B6A7 | |
| Unconjugated | |
| INS | |
| ILPR, INS, Insulin, IRDN | |
| Mouse | |
| Protein A | |
| RUO | |
| 3630 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction