Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Mouse, Clone: SPM139, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2329740.2MG
Description
Insulin Monoclonal antibody specifically detects Antigen in Bovine, Human, Mouse (Negative), Porcine, Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.Specifications
| Insulin | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| 6 kDa | |
| 0.2 mg | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Human, Pig, Bovine, Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM139 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P01308 | |
| INS | |
| Full length (1-84 amino acid) purified pig insulin, conjugated to KLH | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction