Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin/Proinsulin Antibody (D3E7cc), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB359464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359464 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359464 Supplier Novus Biologicals Supplier No. NB200435C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin/Proinsulin Monoclonal antibody specifically detects Insulin/Proinsulin in Human, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA

Specifications

Antigen Insulin/Proinsulin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone D3E7cc
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human insulin.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.