Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin/Proinsulin Antibody (D3E7cc), Novus Biologicals™
SDP

Catalog No. NB200435SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.04 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200435SS 0.04 mg
NB200435 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200435SS Supplier Novus Biologicals Supplier No. NB200435SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin/Proinsulin Monoclonal specifically detects Insulin/Proinsulin in Human, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.

Specifications

Antigen Insulin/Proinsulin
Applications Immunohistochemistry, ELISA
Classification Monoclonal
Clone D3E7
Conjugate Unconjugated
Dilution Immunohistochemistry, Enzyme Immunoassay
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Kd for this is 6.3 x 10^8M.This is specific for both insulin and proinsulin
Purification Method Protein A purified
Quantity 0.04 mg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity This is specific for both insulin and proinsulin
Target Species Human, Rat, Pig, Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.