Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin R/CD220 Antibody (243522) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB399687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399687 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399687 Supplier Novus Biologicals Supplier No. FAB15442A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Insulin R/CD220
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 243522
Conjugate APC
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220, His28-Lys956, Accession # P06213
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3643
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.