Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin R/CD220 Antibody (83-7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB014606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014606 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014606 Supplier Novus Biologicals Supplier No. NBP281039AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Insulin R/CD220
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 83-7
Conjugate Alexa Fluor 647
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Gene Symbols INSR
Host Species Rabbit
Immunogen Epitope within amino acids 140-301 (the cysteine rich region) of the extracellular domain of the human Insulin R/CD220.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3643
Test Specificity Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.