Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin R/CD220 Antibody (INSR/1661), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB384152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384152 Supplier Novus Biologicals Supplier No. NBP308837V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA

Specifications

Antigen Insulin R/CD220
Applications ELISA
Classification Monoclonal
Clone INSR/1661
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Host Species Mouse
Immunogen Recombinant fragment of extracellular domain of human Insulin Receptor alpha (exact sequence is proprietary) (Uniprot: P06213)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3643
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.