Learn More
Description
Specifications
Specifications
| Antigen | Insulin Receptor beta |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR |
| Gene Symbols | INSR |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1332 and 1382 of human Insulin Receptor, beta Subunit using the numbering given in entry NP_000199.2 (GeneID 3643). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
