Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor beta Antibody (C18C4), Novus Biologicals™
SDP

Catalog No. NBP11919205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11919205 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11919205 Supplier Novus Biologicals Supplier No. NBP1191920.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Insulin Receptor beta Monoclonal specifically detects Insulin Receptor beta in Human, Mouse, Rat, Porcine, Bovine, Hamster, Rabbit, Sheep samples. It is validated for Western Blot.

Specifications

Antigen Insulin Receptor beta
Applications Western Blot
Classification Monoclonal
Clone C18C4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Gene Symbols INSR
Host Species Mouse
Immunogen Recombinant cytoplasmic domain of human Insulin Receptor beta
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3643
Test Specificity Insulin Receptor beta (C18C4)
Target Species Human, Mouse, Rat, Pig, Bovine, Hamster, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.