Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ INT 131
GREENER_CHOICE

Catalog No. p-7250119
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PPARγ partial agonist

INT 131 is a potent and selective PPARγ partial agonist (EC50 = 4 nM; Ki = 3.7 nM). It has a 20-fold higher affinity for PPARγ compared with that of Rosiglitazone (Cat. No. 5325). INT 131 improves glucose tolerance in a rodent model of diabetes without inducing hemodynamic and cardiovascular effects. Antidiabetic agent.

Specifications

Chemical Name or Material 2,4-Dichloro-N-[3,5-dichloro-4-(3-quinolinyloxy)phenyl]benzenesulfonamide
CAS 315224-26-1
Quantity 10 mg
Target PPAR gamma/NR1C3
Molecular Formula C21H12Cl4N2O3S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.