Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INT4 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291617 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291617 Supplier Novus Biologicals Supplier No. NB10060660APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INT4 Polyclonal specifically detects INT4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen INT4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias INT4, integrator complex subunit 4, MGC16733, MST093, MSTP093
Host Species Rabbit
Immunogen Maps to a region between residue 913 and 963 of integrator complex subunit 4 using the numbering given in entry NP_291025.3 (GeneID 92105).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92105
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.