Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 1/CD49a Antibody (1090A), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311954 Supplier Novus Biologicals Supplier No. FAB56761PECY55
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha 1/CD49a Monoclonal antibody specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Integrin alpha 1/CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone 1090A
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO derived recombinant human Integrin alpha 1 beta 1, Met1-Pro1141 (Integrin alpha 1) and Gln21-Asp728 (Integrin beta 1), Accession # P56199
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3672
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.