Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 10 Antibody (885501), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345618 Supplier Novus Biologicals Supplier No. FAB9757MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 10 Monoclonal specifically detects Integrin alpha 10 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Integrin alpha 10
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 885501
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias integrin alpha-10, integrin, alpha 10, PRO827
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1, Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1), Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8515
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.