Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2/CD49b Antibody (31H4), FITC, Novus Biologicals™
SDP

Catalog No. NBP250145F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250145F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250145F Supplier Novus Biologicals Supplier No. NBP250145F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen Integrin alpha 2/CD49b
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 31H4
Conjugate FITC
Dilution Flow Cytometry, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2
Gene Symbols ITGA2
Host Species Mouse
Immunogen Purified human b1 integrin preparation from HT1080 fibrosarcoma cell extract.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3673
Test Specificity It is specific to the alpha 2 subunit by relative expression of antigen on various cell lines by Flow Cytomery, and its recognition of affinity purified alpha 2 beta 1 in Dot Blot.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.