Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2/CD49b Antibody (CL7318), Novus Biologicals™
SDP

Catalog No. NB402775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402775 25 μL
NBP276483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB402775 Supplier Novus Biologicals Supplier No. NBP27648325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Integrin alpha 2/CD49b
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL7318
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2
Gene Symbols ITGA2
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ALEAYSETAKVFSIPFHKDCGEDGLCISDLVLDVRQIPAAQEQPFIVSNQNKRLTFSVTLKNKRESAYNTGIVVDFSENLFFASFSLPV
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3673.0
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.