Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (CL7318), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Integrin alpha 2/CD49b |
---|---|
Clone | CL7318 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Integrin alpha 2/CD49b | |
Monoclonal | |
Purified | |
Cellular Markers, Innate Immunity, Signal Transduction | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
3673.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: ALEAYSETAKVFSIPFHKDCGEDGLCISDLVLDVRQIPAAQEQPFIVSNQNKRLTFSVTLKNKRESAYNTGIVVDFSENLFFASFSLPV | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CL7318 | |
Unconjugated | |
Mouse | |
Human | |
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
ITGA2 | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title