Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b beta 3 Antibody (10E5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB015571 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015571 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015571 Supplier Novus Biologicals Supplier No. NBP281080AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Integrin alpha 2b beta 3
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1000000
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Symbols ITGA2B
Host Species Rabbit
Immunogen Human blood platelets.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Test Specificity The antibody binds to Integrin alpha 2b beta 3 with an affinity (Kd) of 16.2+-6 nM. The antibody completely inhibits platelet aggregation at 7.8 ug/ml.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.