Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b beta 3 Antibody (2530A), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346494 Supplier Novus Biologicals Supplier No. FAB76161MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Integrin alpha 2b beta 3
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 2530A
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BDPLT16;BDPLT2;CD41;CD41B;GP2B;GPIIb;GT;GT1;GTA;HPA3;Integrin alpha-IIb;ITGA2B;PPP1R93
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived heterodimer of Human Integrin alpha 2b (Leu32-Arg993, Accession P08514) and Human Integrin beta 3 (Gly27-Asp718, Accession P05106)
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.