Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (745201) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB374390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374390 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374390 Supplier Novus Biologicals Supplier No. FAB7616D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal antibody specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Integrin alpha 2b/CD41
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 745201
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Host Species Mouse
Immunogen heterodimer of human Integrin alpha 2B (Leu32-Arg993; R887L) Accession P08514 + human Integrin beta 3 (Gly27-Asp718) Accession P05106,
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3674
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.