Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (IVA30), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB500496V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500496V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500496V Supplier Novus Biologicals Supplier No. NB500496V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human, Bovine samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone IVA30
Conjugate DyLight 405
Dilution Flow Cytometry, ELISA
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Gene Symbols ITGA2B
Host Species Mouse
Immunogen Bovine trombocytes.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3674
Test Specificity The antibody IVA30 reacts with CD41/CD61 complex (bovine), one of the earliest markers of the megakaryocytic lineage. CD41/CD61 complex acts as the receptor for fibrinogen, fibronectin, Von Willebrand factor, vitronectin, thrombin and mediates platelets aggregation. CD41/CD61 is expressed only by platelets and megakaryocytes; the complex may be absent or strongly reduced in Glanzmann's thrombasthenia (GT).
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.