Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (MWReg30), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP14341A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP14341A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP14341A30 Supplier Novus Biologicals Supplier No. NBP143415AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone MWReg30
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Gene Symbols ITGA2B
Host Species Rat
Immunogen The immunogen for this antibody was CD41.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3674.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.