Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (MWReg30) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP14341505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14341505 0.05 mg
NBP14341501 0.1 mg
NBP1434155 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14341505 Supplier Novus Biologicals Supplier No. NBP1434150.05MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry, ELISA, Immunoprecipitation
Classification Monoclonal
Clone MWReg30
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation PBS, pH 7.2 with 0.09% Sodium Azide
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Gene Symbols ITGA2B
Host Species Rat
Immunogen The immunogen for this antibody was CD41.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.