Learn More
Integrin alpha 2b/CD41 Antibody (MWReg30), DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Integrin alpha 2b/CD41 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | MWReg30 |
| Conjugate | DyLight 550 |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK |
| Gene Symbols | ITGA2B |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.