Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (PM6/248) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017555 0.025 mg
NB10063779 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017555 Supplier Novus Biologicals Supplier No. NB100637790.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry
Classification Monoclonal
Clone PM6/248
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P08514
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Gene Symbols ITGA2B
Host Species Mouse
Immunogen Human platelet plasma membrane
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Test Specificity NB100-63779 recognizes the human CD41 cell surface antigen, a 140kD glycoprotein expressed by platelets and megakaryocytes. CD41 is also known as platelet glycoprotein IIb, and functions as a receptor for fibrinogen, fibronectin and vWF. It has not been established if clone PM6/248 recognizes free CD41 or CD41 only when complexed with CD61. However, antibody binding is reduced in the presence of EDTA suggesting that the epitope recognized is dependent upon an intact CD41/61 complex.
Target Species Human, Mustelid
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.