Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 3/CD49c Antibody (MM0416-2C39), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP211737R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211737R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211737R Supplier Novus Biologicals Supplier No. NBP211737R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 3/CD49c Monoclonal specifically detects Integrin alpha 3/CD49c in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha 3/CD49c
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0416-2C39
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a
Gene Symbols ITGA3
Host Species Mouse
Immunogen Human recombinant Integrin alpha 3/CD49c
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3675
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.