Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (DATK32), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Integrin alpha 4 beta 7/LPAM-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DATK32 |
| Conjugate | DyLight 680 |
| Dilution | Flow Cytometry |
| Gene Alias | 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha |
| Gene Symbols | ITGA4 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was LPAM1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.